AMBH2D6F8BEF Ambeed, Inc.

4-(3,4-Dichlorophenyl)-4-phenylbutanoic acid

  • MDL number MFCD09833046
  • Cas Number : 79560-18-2
  • Molecular Weight : 309.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-120
Purity 95
Flash Point 215.2
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 432.2�0�t 760 mmHg
Product Link https://www.ambeed.com/products/79560-18-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/79560-18-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart