AMBH2D6F8C0D Ambeed, Inc.

3-(4-(Diethylamino)-2-methylphenyl)-3-(1,2-dimethyl-1H-indol-3-yl)isobenzofuran-1(3H)-one

  • MDL number MFCD08276348
  • Cas Number : 36499-49-7
  • Molecular Weight : 438.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/36499-49-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/36499-49-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart