AMBH2D6F8C27 Ambeed, Inc.

Ethyl 2-(imidazo[1,2-a]pyridin-3-yl)acetate

  • MDL number MFCD13183145
  • Cas Number : 101820-69-3
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/101820-69-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/101820-69-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart