AMBH2D6F8C3A Ambeed, Inc.

5,6,7,8-Tetrahydroimidazo[1,2-a]pyridin-6-ol

  • MDL number MFCD21336103
  • Cas Number : 1100750-16-0
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1100750-16-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1100750-16-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart