AMBH2D6F8C58 Ambeed, Inc.

1-(2,6-Diisopropylphenyl)-2-phenyl-1H-imidazole

  • MDL number MFCD27923076
  • Cas Number : 914306-50-6
  • Molecular Weight : 304.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/914306-50-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/914306-50-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart