AMBH2D6F8C60 Ambeed, Inc.

4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine

  • MDL number MFCD06796987
  • Cas Number : 13104-56-8
  • Molecular Weight : 339.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161-163
Purity 97
Flash Point 178.9
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 504.5�t 760 mmHg
Product Link https://www.ambeed.com/products/13104-56-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13104-56-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart