AMBH2D6F8C97 Ambeed, Inc.

Methyl 3-aminooxetane-3-carboxylate

  • MDL number MFCD20922764
  • Cas Number : 1363383-31-6
  • Molecular Weight : 131.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1363383-31-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1363383-31-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart