AMBH2D6F8CB5 Ambeed, Inc.

2,6-Diazaspiro[3.4]octan-7-one oxalate

  • MDL number MFCD27923647
  • Cas Number : 1588440-97-4
  • Molecular Weight : 216.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1588440-97-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1588440-97-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart