AMBH2D6F8CDB Ambeed, Inc.

7-Methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine

  • MDL number MFCD13193598
  • Cas Number : 1260879-61-5
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1260879-61-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1260879-61-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart