AMBH2D6F8CDE Ambeed, Inc.

5-Bromo-2-(methylsulfonyl)pyrimidin-4-amine

  • MDL number MFCD13193626
  • Cas Number : 1379324-53-4
  • Molecular Weight : 252.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 233.4
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 462.4�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1379324-53-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1379324-53-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart