AMBH2D6F8D85 Ambeed, Inc.

2-Chloro-4-cyano-6-fluorobenzoic acid

  • MDL number MFCD22690817
  • Cas Number : 1258298-29-1
  • Molecular Weight : 199.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1258298-29-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1258298-29-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart