AMBH2D6F8DF5 Ambeed, Inc.

2-Chloro-6-methoxy-4-(trifluoromethyl)pyridine

  • MDL number MFCD11845745
  • Cas Number : 1160994-99-9
  • Molecular Weight : 211.5700

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1160994-99-9.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1160994-99-9.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart