AMBH2D6F8E07 Ambeed, Inc.

2-(((Benzyloxy)carbonyl)amino)-3,3,3-trifluoropropanoic acid

  • MDL number MFCD10697679
  • Cas Number : 10068-52-7
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112
Purity 97
Flash Point 196.5
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 401.3�0�t 760 mmHg
Product Link https://www.ambeed.com/products/10068-52-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/10068-52-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart