AMBH2D6F8E50 Ambeed, Inc.

6-Bromo-2,3-dihydro-1H-indene-1-carboxylic acid

  • MDL number MFCD10697921
  • Cas Number : 52651-16-8
  • Molecular Weight : 241.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-145
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 357.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/52651-16-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/52651-16-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart