AMBH2D6F8E66 Ambeed, Inc.

9-(4-Bromophenyl)-10-phenylanthracene

  • MDL number MFCD24842536
  • Cas Number : 625854-02-6
  • Molecular Weight : 409.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/625854-02-6.html

Safety Information

signalword Warning
pcodes P261-P273-P305+P351+P338
MSDS https://www.ambeed.com/products/625854-02-6.html
pictograms GHS07
hstatements H302-H315-H319-H335-H412
Availability details for
Quantity
  • Availability
  • Add to Cart