AMBH2D6F8E73 Ambeed, Inc.

6-Hydroxyazepan-2-one

  • MDL number MFCD15529627
  • Cas Number : 1292369-55-1
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1292369-55-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1292369-55-1.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart