AMBH2D6F8EB1 Ambeed, Inc.

Methyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate

  • MDL number MFCD15203666
  • Cas Number : 943112-78-5
  • Molecular Weight : 190.2000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/943112-78-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/943112-78-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart