AMBH2D6F8EDA Ambeed, Inc.

Imidazo[1,5-a]pyridine-1-carbaldehyde

  • MDL number MFCD06739249
  • Cas Number : 56671-67-1
  • Molecular Weight : 146.1500

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/56671-67-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/56671-67-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart