AMBH2D6F8EDF Ambeed, Inc.

Methyl 7-fluoro-1H-indole-5-carboxylate

  • MDL number MFCD06656840
  • Cas Number : 256935-98-5
  • Molecular Weight : 193.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/256935-98-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/256935-98-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart