AMBH2D6F8F28 Ambeed, Inc.

Methyl 6-chloro-1H-indazole-3-carboxylate

  • MDL number MFCD07371602
  • Cas Number : 717134-47-9
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/717134-47-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/717134-47-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart