AMBH2D6F8F6E Ambeed, Inc.

2-Iodo-N-phenylacetamide

  • MDL number MFCD16170501
  • Cas Number : 7212-28-4
  • Molecular Weight : 261.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7212-28-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7212-28-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart