AMBH2D6F9858 Ambeed, Inc.

6-Methylquinolin-8-ol

  • MDL number MFCD18412519
  • Cas Number : 20984-33-2
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 328.1�0�t 760 mmHg
Product Link https://www.ambeed.com/products/20984-33-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/20984-33-2.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart