AMBH2D6F99EC Ambeed, Inc.

4-Pyrimidin-2-yl-benzoic acid

  • MDL number MFCD01318668
  • Cas Number : 199678-12-1
  • Molecular Weight : 200.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238°C
Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 301.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/199678-12-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/199678-12-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart