AMBH2D6F9A17 Ambeed, Inc.

(1-Methylpiperidin-2-yl)methanamine dihydrochloride

  • MDL number MFCD27996802
  • Cas Number : 1440799-70-1
  • Molecular Weight : 201.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1440799-70-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1440799-70-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart