AMBH2D6F9A3D Ambeed, Inc.

Ethyl 4-hydroxy-2-methylthiazole-5-carboxylate

  • MDL number MFCD08275722
  • Cas Number : 20737-48-8
  • Molecular Weight : 187.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/20737-48-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/20737-48-8.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart