AMBH2D6F9A5B Ambeed, Inc.

6-Bromo-2,3-dihydrobenzofuran-3-ol

  • MDL number MFCD22574142
  • Cas Number : 1392072-52-4
  • Molecular Weight : 215.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1392072-52-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1392072-52-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart