AMBH2D6F9A9A Ambeed, Inc.

2,7-Diazaspiro[3.5]nonan-1-one

  • MDL number MFCD13180578
  • Cas Number : 1147422-92-1
  • Molecular Weight : 140.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 174.5
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 352.9�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1147422-92-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1147422-92-1.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart