AMBH2D6F9B1E Ambeed, Inc.

Methyl 4-chloro-1H-pyrrolo[2,3-b]pyridine-5-carboxylate

  • MDL number MFCD09755979
  • Cas Number : 951625-93-7
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/951625-93-7.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/951625-93-7.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart