AMBH2D6F9BE3 Ambeed, Inc.

4-(tert-Butyl)-2-chloropyridine

  • MDL number MFCD12131129
  • Cas Number : 81167-60-4
  • Molecular Weight : 169.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/81167-60-4.html

Safety Information

signalword Danger
pcodes P261-P280-P301+P310-P311
MSDS https://www.ambeed.com/products/81167-60-4.html
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart