AMBH2D6F9C08 Ambeed, Inc.

1,2,3,4-Tetrahydrobenzofuro[2,3-c]pyridine hydrochloride

  • MDL number MFCD28099338
  • Cas Number : 113411-62-4
  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/113411-62-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/113411-62-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart