AMBH2D6F9C5B Ambeed, Inc.

2-Bromo-2'-chloropropiophenone

  • MDL number MFCD12031558
  • Cas Number : 75815-22-4
  • Molecular Weight : 247.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 110.7
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 259.5�t 760 mmHg
Product Link https://www.ambeed.com/products/75815-22-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/75815-22-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart