AMBH2D6F9C98 Ambeed, Inc.

4-Bromo-5-methylpyridin-2(1H)-one

  • MDL number MFCD16610029
  • Cas Number : 1227578-85-9
  • Molecular Weight : 188.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-5-methylpyridin-2-ol
Purity 95
Flash Point 130.3
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 291.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1227578-85-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1227578-85-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart