AMBH2D6FA2F4 Ambeed, Inc.

Ethyl 5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine-1-carboxylate

  • MDL number MFCD09832125
  • Cas Number : 951627-01-3
  • Molecular Weight : 195.2200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/951627-01-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/951627-01-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart