AMBH2D6FA33A Ambeed, Inc.

5-Oxo-1-phenylpyrrolidine-2-carboxylic acid

  • MDL number MFCD10004175
  • Cas Number : 18133-18-1
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/18133-18-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/18133-18-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart