AMBH2D6FA371 Ambeed, Inc.

(5-Methylbenzofuran-2-yl)boronic acid

  • MDL number MFCD28101768
  • Cas Number : 143381-72-0
  • Molecular Weight : 175.9800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/143381-72-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/143381-72-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart