AMBH2D6FA3C9 Ambeed, Inc.

1-(Cyanomethyl)pyrrolidin-1-ium trifluoromethanesulfonate

  • MDL number MFCD23726589
  • Cas Number : 573987-48-1
  • Molecular Weight : 260.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/573987-48-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/573987-48-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart