AMBH2D6FA3D3 Ambeed, Inc.

N-Cyclopropyl-N-methylpyrrolidin-3-amine

  • MDL number MFCD16709553
  • Cas Number : 250275-00-4
  • Molecular Weight : 140.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/250275-00-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/250275-00-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart