AMBH2D6FA3F6 Ambeed, Inc.

3-Cyano-1H-indazole

  • MDL number MFCD01860240
  • Cas Number : 50264-88-5
  • Molecular Weight : 143.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 126.669
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 370.3�t 760 mmHg
Product Link https://www.ambeed.com/products/50264-88-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/50264-88-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart