AMBH2D6FA46A Ambeed, Inc.

5-(4-Bromophenyl)-4,6-dichloropyrimidine

  • MDL number MFCD13151897
  • Cas Number : 146533-41-7
  • Molecular Weight : 303.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 176.8
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 368.7�0�t 760 mmHg
Product Link https://www.ambeed.com/products/146533-41-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/146533-41-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart