AMBH2D6FA48F Ambeed, Inc.

4-(Difluoromethyl)pyridin-2-amine

  • MDL number MFCD25476528
  • Cas Number : 1346536-47-7
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 106.9°C
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 253.2°35.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/1346536-47-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1346536-47-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart