AMBH2D6FA4A5 Ambeed, Inc.

7-Methylpyrazolo[1,5-a][1,3,5]triazin-4(3H)-one

  • MDL number MFCD12755918
  • Cas Number : 55457-11-9
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/55457-11-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/55457-11-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart