AMBH2D6FA4AF Ambeed, Inc.

7,7-Difluorotetrahydropyrrolo[1,2-a]pyrazine-1,3(2H,4H)-dione

  • MDL number MFCD27501044
  • Cas Number : 1624260-16-7
  • Molecular Weight : 190.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1624260-16-7.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1624260-16-7.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart