AMBH2D6FA4C8 Ambeed, Inc.

4''-(Pentyloxy)-[1,1':4',1''-terphenyl]-4-carboxylic acid

  • MDL number MFCD17019206
  • Cas Number : 158938-08-0
  • Molecular Weight : 360.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/158938-08-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/158938-08-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart