AMBH2D6FA4EF Ambeed, Inc.

Methyl 3-(1,3-dioxoisoindolin-2-yl)-2-hydroxypropanoate

  • MDL number MFCD28134513
  • Cas Number : 153744-36-6
  • Molecular Weight : 249.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/153744-36-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/153744-36-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart