AMBH2D6FA578 Ambeed, Inc.

Methyl 3-((2,2-dimethylbutanoyl)thio)propanoate

  • MDL number MFCD22200648
  • Cas Number : 938063-63-9
  • Molecular Weight : 218.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/938063-63-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/938063-63-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart