AMBH2D6FA61D Ambeed, Inc.

Benzene-1,3,5-triamine xhydrochloride

  • MDL number N/A
  • Cas Number : 88010-00-8
  • Molecular Weight : 278.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/88010-00-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/88010-00-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart