AMBH2D6FA663 Ambeed, Inc.

7H-Benzo[de]anthracen-7-one

  • MDL number MFCD00003585
  • Cas Number : 82-05-3
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170°C
Purity 98
Flash Point 196.1°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 436.2°C at 760 mmHg
Product Link https://www.ambeed.com/products/82-05-3.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/82-05-3.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart