AMBH2D6FA67F Ambeed, Inc.

Rucaparib phosphate

  • MDL number MFCD17010269
  • Cas Number : 459868-92-9
  • Molecular Weight : 421.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Fluoro-2-(4-((methylamino)methyl)phenyl)-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one phosphate
Purity 99
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/459868-92-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/459868-92-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart