AMBH2D6FA702 Ambeed, Inc.

3-Bromo-2-chloro-5-nitropyridin-4-amine

  • MDL number MFCD18632701
  • Cas Number : 1268521-33-0
  • Molecular Weight : 252.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1268521-33-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1268521-33-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart