AMBH2D6FB045 Ambeed, Inc.

2-(3,5-Bis(trifluoromethyl)phenyl)-2-methylpropanoic acid

  • MDL number MFCD09032572
  • Cas Number : 289686-70-0
  • Molecular Weight : 300.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/289686-70-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/289686-70-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart